siRNA Details
| Gene Symbol: Steap4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044265.1 | NP_001037730.1 | 2, 3 | 575 | ||
| GenBank mRNA | AY325154.1 | - | 487 | |||
| BC097353.1 | - | 2, 3 | 575 | 7387686 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Steap4| Gene Symbol: Steap4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044265.1 | NP_001037730.1 | 2, 3 | 575 | ||
| GenBank mRNA | AY325154.1 | - | 487 | |||
| BC097353.1 | - | 2, 3 | 575 | 7387686 | ||