siRNA Details
Gene Symbol: Phf14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110492.1 | NP_001103962.1 | 14 | 2790 | ||
XM_006236104.2 | XP_006236166.1 | 14 | 2826 | |||
XM_006236105.2 | XP_006236167.1 | 14 | 2565 | |||
XM_006236107.2 | XP_006236169.1 | 14 | 2826 | |||
XM_006236108.2 | XP_006236170.1 | 14 | 2826 | |||
XM_006236109.2 | XP_006236171.1 | 14 | 2826 | |||
XM_006236111.2 | XP_006236173.1 | 14 | 2825 | |||
XM_008762716.1 | XP_008760938.1 | 14 | 2826 | |||
GenBank mRNA | BC166456.1 | - | 14 | 2521 | 9025134 | |
FQ209867.1 | - | 14 | 2754 | |||
FQ221829.1 | - | 13 | 3068 |
