siRNA Details
| Gene Symbol: Thsd7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191970.1 | NP_001178899.1 | 8 | 2102 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Thsd7a| Gene Symbol: Thsd7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191970.1 | NP_001178899.1 | 8 | 2102 | ||