siRNA Details
| Gene Symbol: LOC500034 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109221.2 | NP_001102691.1 | 2, 3 | 286 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC500034| Gene Symbol: LOC500034 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109221.2 | NP_001102691.1 | 2, 3 | 286 | ||