siRNA Details
Gene Symbol: Ppp1r3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109222.1 | NP_001102692.1 | 4 | 1271 | ||
XM_008762717.1 | XP_008760939.1 | 4 | 1313 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ppp1r3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109222.1 | NP_001102692.1 | 4 | 1271 | ||
XM_008762717.1 | XP_008760939.1 | 4 | 1313 |