siRNA Details
Gene Symbol: Foxp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271104.1 | NP_001258033.1 | 5 | 843 | ||
XM_002729285.4 | XP_002729331.1 | 3 | 653 | |||
XM_006236115.2 | XP_006236177.1 | 3 | 1326 | |||
XM_008762718.1 | XP_008760940.1 | 3 | 411 | |||
XM_008762719.1 | XP_008760941.1 | 3 | 653 | |||
XM_008762720.1 | XP_008760942.1 | 3 | 411 | |||
XM_008762721.1 | XP_008760943.1 | 3 | 653 |
