siRNA Details
| Gene Symbol: Tes | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039344.2 | NP_001034433.1 | 3 | 345 | ||
| GenBank mRNA | BC129069.1 | - | 3 | 345 | 7100907 | |
| DQ339047.1 | - | 3 | 346 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tes| Gene Symbol: Tes | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039344.2 | NP_001034433.1 | 3 | 345 | ||
| GenBank mRNA | BC129069.1 | - | 3 | 345 | 7100907 | |
| DQ339047.1 | - | 3 | 346 | |||