siRNA Details
| Gene Symbol: Cped1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002729287.4 | XP_002729333.2 | 6 | 1738 | ||
| GenBank mRNA | BC100097.1 | - | 6 | 1029 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cped1| Gene Symbol: Cped1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002729287.4 | XP_002729333.2 | 6 | 1738 | ||
| GenBank mRNA | BC100097.1 | - | 6 | 1029 | ||