siRNA Details
| Gene Symbol: Zfp800 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109225.1 | NP_001102695.1 | 4 | 1459 | ||
| XM_008762812.1 | XP_008761034.1 | 5 | 1479 | |||
| XM_008762813.1 | XP_008761035.1 | 4 | 1460 | |||
| XM_008762814.1 | XP_008761036.1 | 5 | 1624 | |||
| XM_008762815.1 | XP_008761037.1 | 5 | 1700 | |||
| XM_008762816.1 | XP_008761038.1 | 5 | 1479 | |||

Zfp800
Genomic Map