siRNA Details
| Gene Symbol: Zfp800 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109225.1 | NP_001102695.1 | 2 | 188 | ||
| XM_008762812.1 | XP_008761034.1 | 3 | 208 | |||
| XM_008762813.1 | XP_008761035.1 | 2 | 189 | |||
| XM_008762814.1 | XP_008761036.1 | 3 | 353 | |||
| XM_008762815.1 | XP_008761037.1 | 3 | 429 | |||
| XM_008762816.1 | XP_008761038.1 | 3 | 208 | |||

Zfp800
Genomic Map