siRNA Details
Gene Symbol: Fam71f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024324.1 | NP_001019495.1 | 2 | 312 | ||
XM_006236223.2 | XP_006236285.1 | 4 | 674 | |||
XM_006236224.2 | XP_006236286.1 | 4 | 674 | |||
XM_006236225.2 | XP_006236287.1 | 4 | 674 | |||
XM_008762818.1 | XP_008761040.1 | 4 | 674 | |||
GenBank mRNA | BC079297.1 | - | 2 | 312 | 7134497 |
