siRNA Details
| Gene Symbol: Tspan33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109227.1 | NP_001102697.1 | 7 | 905 | ||
| XM_006236227.2 | XP_006236289.1 | 7 | 891 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tspan33| Gene Symbol: Tspan33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109227.1 | NP_001102697.1 | 7 | 905 | ||
| XM_006236227.2 | XP_006236289.1 | 7 | 891 | |||