siRNA Details
Gene Symbol: Ssmem1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109228.1 | NP_001102698.1 | 3 | 721 | ||
XM_006236228.2 | XP_006236290.1 | 3 | 673 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ssmem1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109228.1 | NP_001102698.1 | 3 | 721 | ||
XM_006236228.2 | XP_006236290.1 | 3 | 673 |