siRNA Details
| Gene Symbol: LOC500077 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024325.1 | NP_001019496.1 | 2 | 524 | ||
| GenBank mRNA | BC079043.1 | - | 2 | 524 | 7114933 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC500077| Gene Symbol: LOC500077 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024325.1 | NP_001019496.1 | 2 | 524 | ||
| GenBank mRNA | BC079043.1 | - | 2 | 524 | 7114933 | |