siRNA Details
Gene Symbol: Stra8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006236282.2 | XP_006236344.1 | 5 | 2860 | ||
XM_575429.4 | XP_575429.2 | 4 | 603 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Stra8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006236282.2 | XP_006236344.1 | 5 | 2860 | ||
XM_575429.4 | XP_575429.2 | 4 | 603 |