siRNA Details
| Gene Symbol: Rab19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024326.1 | NP_001019497.1 | 4 | 652 | ||
| XM_008762820.1 | XP_008761042.1 | 4 | 1080 | |||
| GenBank mRNA | BC088443.1 | - | 4 | 652 | 7126386 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab19| Gene Symbol: Rab19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024326.1 | NP_001019497.1 | 4 | 652 | ||
| XM_008762820.1 | XP_008761042.1 | 4 | 1080 | |||
| GenBank mRNA | BC088443.1 | - | 4 | 652 | 7126386 | |