siRNA Details
| Gene Symbol: Tas2r144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025150.1 | NP_001020321.1 | 1 | 231 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tas2r144| Gene Symbol: Tas2r144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025150.1 | NP_001020321.1 | 1 | 231 | ||