siRNA Details
| Gene Symbol: Fam131b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025046.1 | NP_001020217.2 | 5, 6 | 586 | ||
| XM_008762903.1 | XP_008761125.1 | 2, 3 | 238 | |||
| GenBank mRNA | BC092642.1 | - | 5, 6 | 586 | 7318030 | |

Fam131b
Genomic Map