siRNA Details
| Gene Symbol: Zfp398 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109230.1 | NP_001102700.1 | 3, 4 | 777 | ||
| XM_008762904.1 | XP_008761126.1 | 2, 3 | 386 | |||
| XM_008762905.1 | XP_008761127.1 | 2, 3 | 268 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp398| Gene Symbol: Zfp398 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109230.1 | NP_001102700.1 | 3, 4 | 777 | ||
| XM_008762904.1 | XP_008761126.1 | 2, 3 | 386 | |||
| XM_008762905.1 | XP_008761127.1 | 2, 3 | 268 | |||