siRNA Details
| Gene Symbol: Zfp467 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024327.1 | NP_001019498.1 | 4 | 678 | ||
| XM_006236431.2 | XP_006236493.1 | 5 | 763 | |||
| XM_006236432.2 | XP_006236494.1 | 5 | 1013 | |||
| XM_006236433.2 | XP_006236495.1 | 5 | 1294 | |||
| XM_006236434.2 | XP_006236496.1 | 5 | 871 | |||
| GenBank mRNA | BC086534.1 | - | 4 | 678 | 7097755 | |

Zfp467
Genomic Map