siRNA Details
| Gene Symbol: Gimap8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033923.1 | NP_001029095.1 | 5 | 2158 | ||
| XM_006236441.2 | XP_006236503.1 | 6 | 2135 | |||
| GenBank mRNA | AJ633685.1 | - | 5 | 1999 | ||
| BC099228.1 | - | 5 | 2158 | 7389353 | ||
| DQ125335.1 | - | 5 | 2188 | |||
| DQ125336.1 | - | 5 | 2188 | |||

Gimap8
Genomic Map