siRNA Details
| Gene Symbol: Gimap7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024328.2 | NP_001019499.1 | 2 | 448 | ||
| GenBank mRNA | AJ633682.1 | - | 887 | |||
| BC091210.1 | - | 2 | 394 | 7381387 | ||
| CV075794.1 | - | 2 | 448 | |||
| DQ125348.1 | - | 2 | 471 | |||
| DQ125349.1 | - | 2 | 471 | |||
| FQ232730.1 | - | 2 | 479 | |||

Gimap7
Genomic Map