siRNA Details
| Gene Symbol: Ccdc126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109232.2 | NP_001102702.1 | 2 | 292 | ||
| GenBank mRNA | BC168861.1 | - | 2 | 292 | 7129053 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc126| Gene Symbol: Ccdc126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109232.2 | NP_001102702.1 | 2 | 292 | ||
| GenBank mRNA | BC168861.1 | - | 2 | 292 | 7129053 | |