siRNA Details
| Gene Symbol: Fam221a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025771.2 | NP_001020942.1 | 6 | 800 | ||
| XM_006236467.2 | XP_006236529.1 | 6 | 773 | |||
| XM_006236468.2 | XP_006236530.1 | 5 | 787 | |||
| GenBank mRNA | BC097434.1 | - | 6 | 801 | 7455430 | |

Fam221a
Genomic Map