siRNA Details
| Gene Symbol: Stk31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191971.1 | NP_001178900.1 | 19 | 2392 | ||
| XM_008762906.1 | XP_008761128.1 | 19 | 2419 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stk31| Gene Symbol: Stk31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191971.1 | NP_001178900.1 | 19 | 2392 | ||
| XM_008762906.1 | XP_008761128.1 | 19 | 2419 | |||