siRNA Details
| Gene Symbol: LOC500124 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024329.1 | NP_001019500.1 | 2 | 201 | ||
| GenBank mRNA | BC078992.1 | - | 2 | 201 | 7112600 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC500124| Gene Symbol: LOC500124 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024329.1 | NP_001019500.1 | 2 | 201 | ||
| GenBank mRNA | BC078992.1 | - | 2 | 201 | 7112600 | |