siRNA Details
| Gene Symbol: RGD1564419 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109234.1 | NP_001102704.1 | 3, 4 | 1129 | ||
| XM_006236504.2 | XP_006236566.1 | 1, 2 | 855 | |||
| GenBank mRNA | BC169060.1 | - | 1, 2 | 817 | 7930294 | |

RGD1564419
Genomic Map