siRNA Details
| Gene Symbol: RGD1564419 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109234.1 | NP_001102704.1 | 4 | 1339 | ||
| XM_006236504.2 | XP_006236566.1 | 2 | 1065 | |||
| GenBank mRNA | BC169060.1 | - | 2 | 1027 | 7930294 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1564419| Gene Symbol: RGD1564419 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109234.1 | NP_001102704.1 | 4 | 1339 | ||
| XM_006236504.2 | XP_006236566.1 | 2 | 1065 | |||
| GenBank mRNA | BC169060.1 | - | 2 | 1027 | 7930294 | |