siRNA Details
| Gene Symbol: Plekha8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109235.2 | NP_001102705.1 | 5, 6 | 768 | ||
| XM_006236506.2 | XP_006236568.1 | 5, 6 | 853 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plekha8| Gene Symbol: Plekha8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109235.2 | NP_001102705.1 | 5, 6 | 768 | ||
| XM_006236506.2 | XP_006236568.1 | 5, 6 | 853 | |||