siRNA Details
| Gene Symbol: LOC500148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC079164.1 | - | 2 | 566 | 7129532 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC500148| Gene Symbol: LOC500148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC079164.1 | - | 2 | 566 | 7129532 | |