siRNA Details
| Gene Symbol: Mmrn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001071128.5 | XP_001071128.2 | 7 | 1441 | ||
| XM_006236596.2 | XP_006236658.1 | 7 | 1441 | |||
| XM_006236597.2 | XP_006236659.1 | 8 | 1439 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mmrn1| Gene Symbol: Mmrn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001071128.5 | XP_001071128.2 | 7 | 1441 | ||
| XM_006236596.2 | XP_006236658.1 | 7 | 1441 | |||
| XM_006236597.2 | XP_006236659.1 | 8 | 1439 | |||