siRNA Details
Gene Symbol: Ptcd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134718.1 | NP_001128190.1 | 6 | 400 | ||
XM_008762995.1 | XP_008761217.1 | 9 | 964 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ptcd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134718.1 | NP_001128190.1 | 6 | 400 | ||
XM_008762995.1 | XP_008761217.1 | 9 | 964 |