siRNA Details
Gene Symbol: Lrrtm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134746.1 | NP_001128218.1 | 1 | 1368 | ||
XM_008763024.1 | XP_008761246.1 | 2 | 1687 | |||
XM_008763025.1 | XP_008761247.1 | 3 | 1840 | |||
XM_008763026.1 | XP_008761248.1 | 3 | 1751 | |||
GenBank mRNA | BC168219.1 | - | 1 | 1368 | 7319359 |
