siRNA Details
| Gene Symbol: Aak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173450.1 | NP_001166921.1 | 4 | 963 | ||
| XM_006236833.2 | XP_006236895.1 | 5 | 813 | |||
| XM_006236834.2 | XP_006236896.1 | 5 | 813 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aak1| Gene Symbol: Aak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173450.1 | NP_001166921.1 | 4 | 963 | ||
| XM_006236833.2 | XP_006236895.1 | 5 | 813 | |||
| XM_006236834.2 | XP_006236896.1 | 5 | 813 | |||