siRNA Details
| Gene Symbol: Arhgap25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109247.1 | NP_001102717.1 | 4 | 801 | ||
| XM_006236838.2 | XP_006236900.1 | 4 | 2960 | |||
| XM_006236839.2 | XP_006236901.1 | 3 | 656 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arhgap25| Gene Symbol: Arhgap25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109247.1 | NP_001102717.1 | 4 | 801 | ||
| XM_006236838.2 | XP_006236900.1 | 4 | 2960 | |||
| XM_006236839.2 | XP_006236901.1 | 3 | 656 | |||