siRNA Details
| Gene Symbol: Rab43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024331.1 | NP_001019502.1 | 3 | 626 | ||
| GenBank mRNA | AY593256.1 | - | 3 | 626 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab43| Gene Symbol: Rab43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024331.1 | NP_001019502.1 | 3 | 626 | ||
| GenBank mRNA | AY593256.1 | - | 3 | 626 | ||