siRNA Details
| Gene Symbol: Chst13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191974.1 | NP_001178903.1 | 3 | 315 | ||
| XM_006236870.1 | XP_006236932.1 | 2 | 216 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chst13| Gene Symbol: Chst13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191974.1 | NP_001178903.1 | 3 | 315 | ||
| XM_006236870.1 | XP_006236932.1 | 2 | 216 | |||