siRNA Details
| Gene Symbol: RGD1560289 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134623.2 | NP_001128095.2 | 13 | 2265 | ||
| XM_006236917.2 | XP_006236979.1 | 13 | 3581 | |||
| XM_008763127.1 | XP_008761349.1 | 12 | 5306 | |||
| XM_008763128.1 | XP_008761350.1 | 13 | 3886 | |||
| XM_008763129.1 | XP_008761351.1 | 11 | 5297 | |||
| XM_008763130.1 | XP_008761352.1 | 12 | 5307 | |||

RGD1560289
Genomic Map