siRNA Details
| Gene Symbol: Kbtbd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109250.3 | NP_001102720.2 | 3 | 1185 | ||
| XM_006236932.1 | XP_006236994.1 | 3 | 1182 | |||
| GenBank mRNA | BC160902.1 | - | 3 | 1185 | 7454217 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kbtbd8| Gene Symbol: Kbtbd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109250.3 | NP_001102720.2 | 3 | 1185 | ||
| XM_006236932.1 | XP_006236994.1 | 3 | 1182 | |||
| GenBank mRNA | BC160902.1 | - | 3 | 1185 | 7454217 | |