siRNA Details
| Gene Symbol: Ier3ip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047956.1 | NP_001041421.1 | 4 | 338 | ||
| XM_008763163.1 | XP_008761385.1 | 3 | 333 | |||
| GenBank mRNA | AY325263.1 | - | 4 | 338 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ier3ip1| Gene Symbol: Ier3ip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047956.1 | NP_001041421.1 | 4 | 338 | ||
| XM_008763163.1 | XP_008761385.1 | 3 | 333 | |||
| GenBank mRNA | AY325263.1 | - | 4 | 338 | ||