siRNA Details
| Gene Symbol: Cidec | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024333.2 | NP_001019504.2 | 5, 6 | 597 | ||
| NM_001244797.1 | NP_001231726.1 | 5, 6 | 609 | |||
| NM_001244798.1 | NP_001231727.1 | 5, 6 | 598 | |||
| GenBank mRNA | BC083860.1 | - | 5, 6 | 597 | 7126532 | |
| FM043477.1 | - | 5, 6 | 606 | |||

Cidec
Genomic Map