siRNA Details
| Gene Symbol: Fancd2os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109251.1 | NP_001102721.1 | 3 | 570 | ||
| XM_006237051.2 | XP_006237113.1 | 2 | 642 | |||
| XM_006237052.2 | XP_006237114.1 | 2 | 1243 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fancd2os| Gene Symbol: Fancd2os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109251.1 | NP_001102721.1 | 3 | 570 | ||
| XM_006237051.2 | XP_006237113.1 | 2 | 642 | |||
| XM_006237052.2 | XP_006237114.1 | 2 | 1243 | |||