siRNA Details
| Gene Symbol: Zfp248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002729406.4 | XP_002729452.2 | 4 | 1642 | ||
| XM_006237213.2 | XP_006237275.1 | 5 | 1685 | |||
| XM_006237215.2 | XP_006237277.1 | 4 | 1638 | |||
| XM_008763270.1 | XP_008761492.1 | 3 | 1857 | |||
| GenBank mRNA | AY724530.1 | - | 897 | |||

Zfp248
Genomic Map