siRNA Details
| Gene Symbol: Dcp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749821.3 | XP_003749869.1 | 9 | 2065 | ||
| XM_008763272.1 | XP_008761494.1 | 9 | 2233 | |||
| XM_575654.6 | XP_575654.3 | 9 | 2164 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dcp1b| Gene Symbol: Dcp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749821.3 | XP_003749869.1 | 9 | 2065 | ||
| XM_008763272.1 | XP_008761494.1 | 9 | 2233 | |||
| XM_575654.6 | XP_575654.3 | 9 | 2164 | |||