siRNA Details
| Gene Symbol: B4galnt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191976.1 | NP_001178905.1 | 16 | 2457 | ||
| XM_006237267.2 | XP_006237329.1 | 16 | 2760 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

B4galnt3| Gene Symbol: B4galnt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191976.1 | NP_001178905.1 | 16 | 2457 | ||
| XM_006237267.2 | XP_006237329.1 | 16 | 2760 | |||