siRNA Details
| Gene Symbol: Gdf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109671.1 | NP_001103141.1 | 1 | 367 | ||
| GenBank mRNA | DQ372084.1 | - | 351 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gdf3| Gene Symbol: Gdf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109671.1 | NP_001103141.1 | 1 | 367 | ||
| GenBank mRNA | DQ372084.1 | - | 351 | |||