siRNA Details
| Gene Symbol: Acrbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025049.1 | NP_001020220.1 | 2 | 150 | ||
| XM_006237379.2 | XP_006237441.1 | 2 | 690 | |||
| GenBank mRNA | BC079212.1 | - | 2 | 150 | 7131828 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acrbp| Gene Symbol: Acrbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025049.1 | NP_001020220.1 | 2 | 150 | ||
| XM_006237379.2 | XP_006237441.1 | 2 | 690 | |||
| GenBank mRNA | BC079212.1 | - | 2 | 150 | 7131828 | |