siRNA Details
| Gene Symbol: RGD1564770 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001194985.1 | NP_001181914.1 | 1, 2 | 50 | ||
| GenBank mRNA | BC158621.1 | - | 258 | |||
| GU357487.1 | - | 1, 2 | 50 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1564770| Gene Symbol: RGD1564770 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001194985.1 | NP_001181914.1 | 1, 2 | 50 | ||
| GenBank mRNA | BC158621.1 | - | 258 | |||
| GU357487.1 | - | 1, 2 | 50 | |||