siRNA Details
| Gene Symbol: Clec1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109253.1 | NP_001102723.1 | 6 | 1006 | ||
| GenBank mRNA | GU357481.1 | - | 6 | 721 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clec1a| Gene Symbol: Clec1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109253.1 | NP_001102723.1 | 6 | 1006 | ||
| GenBank mRNA | GU357481.1 | - | 6 | 721 | ||