siRNA Details
| Gene Symbol: LOC500350 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047957.1 | NP_001041422.1 | 7 | 1170 | ||
| GenBank mRNA | AY539890.1 | - | 7 | 1170 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC500350| Gene Symbol: LOC500350 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047957.1 | NP_001041422.1 | 7 | 1170 | ||
| GenBank mRNA | AY539890.1 | - | 7 | 1170 | ||